Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pericentrin Antibody, Novus Biologicals™
SDP

Catalog No. NB10063208 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10063208 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10063208 Supplier Novus Biologicals Supplier No. NB10063208
Add to Cart
Add to Cart

Goat Polyclonal Antibody

Pericentrin Polyclonal specifically detects Pericentrin in Human samples. It is validated for ELISA, Western Blot.

Specifications

Antigen Pericentrin
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1 - 3 ug/mL, ELISA >1:32000
Formulation TBS and 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. Q9BW27
Gene Alias Kendrin, KIAA0402, MOPD2, PCN, PCNT2pericentrin B, PCNTB, pericentrin, pericentrin 2 (kendrin), pericentrin, kendrin (KIAA0402)10KENPCTN2, pericentrin-2, pericentrin-380, pericentrin-B, SCKL4kendrin
Gene Symbols PCNT
Host Species Goat
Immunogen Peptide sequence C-ARAIIREGSLEGS corresponding to the C-terminal of pericentrin 1 (NP_079120).
Purification Method Affinity Purified
Quantity 0.05 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5116
Test Specificity NB100-63208 recognizes the C-terminal of human pericentrin 1, a 75 kDa component of the filamentous matrix of the centrosome, expressed in all tissues. When associated with centrosome and Golgi localized protein kinase N-associated protein (CG-NAP), pericentrin 1 anchors the gamma-tubulin ring complex, providing a site for microtubule nucleation and the initial establishment of organized microtubule arrays. It binds calmodulin and is a constituent of nuclear pore proteins (nup) 107-160, a sub-complex of the nuclear pore complex. Pericentrin 1 is thought to play a role in the functioning of the centrosomes, cytoskeleton, and cell-cycle progression.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.