Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pericentrin Antibody, Novus Biologicals™
SDP

Catalog No. NB10061071 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10061071 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10061071 Supplier Novus Biologicals Supplier No. NB10061071
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 7 publications

Pericentrin Polyclonal specifically detects Pericentrin in Human samples. It is validated for Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.

Specifications

Antigen Pericentrin
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:2000 - 1:10000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:1000 - 1:5000, Immunoprecipitation 2 - 10 ug/mg lysate, Immunohistochemistry-Paraffin 1:10 - 1:500
Formulation Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
Gene Accession No. O95613
Gene Alias Kendrin, KIAA0402, MOPD2, PCN, PCNT2pericentrin B, PCNTB, pericentrin, pericentrin 2 (kendrin), pericentrin, kendrin (KIAA0402)10KENPCTN2, pericentrin-2, pericentrin-380, pericentrin-B, SCKL4kendrin
Gene Symbols PCNT
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 175 and 225 of human Pericentrin/Kendrin using the numbering given in entry NP_006022.3 (GeneID 5116).
Molecular Weight of Antigen 378 kDa
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Cellular Markers, Centrosome Markers
Primary or Secondary Primary
Gene ID (Entrez) 5116
Test Specificity Pericentrin/Kendrin - Centrosome Marker.
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.