Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco™ Peridotite Specimen (Igneous Rock)
SCI_ED

Catalog No. p-7076853 Shop All Eisco Products
Click to view available options
Quantity:
12/Pk.
Each

For use in comparing with other igneous rocks, particularly as part of lessons on Bowen's reaction series. EISCO Peridotite Specimen (Igneous Rock) is iron-rich ultramafic rock, with chemistry that suggests genesis from point deep within crust.

  • Useful as classroom demonstration of what is believed to make up most of earth's mantle, far below crust
  • For use in classroom identification labs and exercises
  • Correlates well to Next Generation Science Standards Evidence Statements
  • Contains abundant heavy elements such as iron and magnesium
  • Large crystals indicate slow cooling in magma chamber
  • Comes in clear labeled and barcoded bag for storage

Specifications

Quantity Each
For Use With (Application) Comparing with other igneous rocks, particularly as part of lessons on Bowen's reaction series
Product Type Peridotite Study Specimen (Igneous Rock)
Width (English) Approx. 1 in.
Width (Metric) Approx. 3 cm
Packaging Clear labeled and barcoded bag
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.