Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Perilipin-2/ADFP Antibody (ADFP/1366), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP253405100UG
Description
Perilipin-2/ADFP Monoclonal specifically detects Perilipin-2/ADFP in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Protein Array.Specifications
Perilipin-2/ADFP | |
Monoclonal | |
0.2 mg/mL | |
ELISA 1:100 - 1:2000, Immunocytochemistry/Immunofluorescence 0.5 - 1 ug/ml, Immunohistochemistry-Paraffin 0.5 - 1.0 ug/ml, SDS-Page | |
ADFP, Adipophilin, Adipose differentiation-related proteinadipophilin, ADRP, MGC10598, perilipin 2, perilipin-2 | |
Mouse | |
48 kDa | |
100 μg | |
Cancer, Hypoxia, Lipid and Metabolism, Lipid Droplets, Signal Transduction | |
123 | |
Store at 4C. | |
IgG2b κ |
ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), SDS-Page | |
ADFP/1366 | |
Unconjugated | |
10mM PBS with 0.05% BSA with 0.05% Sodium Azide | |
PLIN2 | |
Recombinant human Adipophilin protein fragment (aa249-376) (exact sequence is proprietary) | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction