Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Perilipin-2/ADFP Antibody (ADFP/1366), Novus Biologicals™

Mouse Monoclonal Antibody
$268.00 - $524.00
Specifications
Antigen | Perilipin-2/ADFP |
---|---|
Clone | ADFP/1366 |
Concentration | 0.2 mg/mL |
Dilution | ELISA 1:100 - 1:2000, Immunocytochemistry/Immunofluorescence 0.5 - 1 ug/ml, Immunohistochemistry-Paraffin 0.5 - 1.0 ug/ml, SDS-Page |
Applications | ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), SDS-Page |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB25340520
![]() |
Novus Biologicals
NBP25340520UG |
20 μg |
Each for $268.00
|
|
|||||
NB253405100
![]() |
Novus Biologicals
NBP253405100UG |
100 μg |
Each for $524.00
|
|
|||||
NB253405200
![]() |
Novus Biologicals
NBP253405200UG |
200 μg | N/A | N/A | N/A | ||||
Description
Perilipin-2/ADFP Monoclonal specifically detects Perilipin-2/ADFP in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Protein Array.Specifications
Perilipin-2/ADFP | |
0.2 mg/mL | |
ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), SDS-Page | |
Unconjugated | |
Mouse | |
Cancer, Hypoxia, Lipid and Metabolism, Lipid Droplets, Signal Transduction | |
10mM PBS with 0.05% BSA with 0.05% Sodium Azide | |
123 | |
Recombinant human Adipophilin protein fragment (aa249-376) (exact sequence is proprietary) | |
Primary | |
Store at 4C. |
ADFP/1366 | |
ELISA 1:100 - 1:2000, Immunocytochemistry/Immunofluorescence 0.5 - 1 ug/ml, Immunohistochemistry-Paraffin 0.5 - 1.0 ug/ml, SDS-Page | |
Monoclonal | |
Purified | |
RUO | |
Human | |
ADFP, Adipophilin, Adipose differentiation-related proteinadipophilin, ADRP, MGC10598, perilipin 2, perilipin-2 | |
PLIN2 | |
IgG2b κ | |
Protein A or G purified | |
48 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title