Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Perilipin-2/ADFP Antibody (ADFP/1493), Alexa Fluor™ 647, Novus Biologicals™
SDP

Catalog No. NB007982 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB007982 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB007982 Supplier Novus Biologicals Supplier No. NBP254312AF647
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Perilipin-2/ADFP Monoclonal specifically detects Perilipin-2/ADFP in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Perilipin-2/ADFP
Applications ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone ADFP/1493
Conjugate Alexa Fluor 647
Dilution ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias ADFP, Adipophilin, Adipose differentiation-related proteinadipophilin, ADRP, MGC10598, perilipin 2, perilipin-2
Gene Symbols PLIN2
Host Species Mouse
Immunogen Recombinant fragment (around aa 249-376) of human Perilipin-2/ADFP protein (exact sequence is proprietary) (Uniprot: Q99541)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Hypoxia, Lipid and Metabolism, Lipid Droplets, Signal Transduction
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG2b λ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.