Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Perilipin-2/ADFP Antibody, DyLight 550
SDP

Catalog No. NB11040878R Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB11040878R 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB11040878R Supplier Novus Biologicals Supplier No. NB11040878R

Rabbit Polyclonal Antibody

Perilipin-2/ADFP Polyclonal specifically detects Perilipin-2/ADFP in Human, Mouse, Porcine, Monkey samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Perilipin-2/ADFP
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 550
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Accession No. Q99541
Gene Alias ADFP, Adipophilin, Adipose differentiation-related proteinadipophilin, ADRP, MGC10598, perilipin 2, perilipin-2
Gene Symbols PLIN2
Host Species Rabbit
Immunogen A synthetic peptide made to an internal portion of human ADFP (within residues 150-250) [Swiss-Prot# Q99541]
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Lipid and Metabolism, Lipid Droplets, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 123
Test Specificity Human and mouse. Immunogen has 100% identity to monkey, porcine and bovine ADFP, 94% identity tp sheep and 84% identity to rat.
Target Species Human, Mouse, Pig, Monkey
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.