Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Perilipin-3/TIP47 Antibody, DyLight 405, Novus Biologicals™
SDP

Catalog No. NB11040764V Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB11040764V 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB11040764V Supplier Novus Biologicals Supplier No. NB11040764V
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Perilipin-3/TIP47 Polyclonal specifically detects Perilipin-3/TIP47 in Human, Mouse, Porcine, Primate samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Perilipin-3/TIP47
Applications Western Blot, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Conjugate DyLight 405
Dilution Western Blot, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Accession No. O60664
Gene Alias Cargo selection protein TIP47, M6PRBP1MGC2012, Mannose-6-phosphate receptor-binding protein 1, perilipin 3,47 kDa MPR-binding protein, perilipin-3, Placental protein 17,47 kDa mannose 6-phosphate receptor-binding protein, PP17MGC11117, tail-interacting protein, 47 kD, TIP47mannose-6-phosphate receptor binding protein 1
Gene Symbols PLIN3
Host Species Rabbit
Immunogen A synthetic peptide made to a region within the C-terminus (within residues 350-435) of the human TIP47 protein. [Swiss-Prot# O60664]
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Diabetes Research, Golgi Apparatus Markers, Lipid and Metabolism, Lipid Droplets, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 10226
Target Species Human, Mouse, Pig, Primate
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.