Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Perilipin-3/TIP47 Antibody, FITC, Novus Biologicals™
SDP

Catalog No. NB052171 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB052171 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB052171 Supplier Novus Biologicals Supplier No. NBP298075F
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Perilipin-3/TIP47 Polyclonal specifically detects Perilipin-3/TIP47 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Perilipin-3/TIP47
Applications Immunofluorescence
Classification Polyclonal
Conjugate FITC
Dilution Immunocytochemistry/Immunofluorescence
Formulation PBS
Gene Alias Cargo selection protein TIP47, M6PRBP1MGC2012, Mannose-6-phosphate receptor-binding protein 1, perilipin 3,47 kDa MPR-binding protein, perilipin-3, Placental protein 17,47 kDa mannose 6-phosphate receptor-binding protein, PP17MGC11117, tail-interacting protein, 47 kD, TIP47mannose-6-phosphate receptor binding protein 1
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human Perilipin-3/TIP47 fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Diabetes Research, Golgi Apparatus Markers, Lipid and Metabolism, Lipid Droplets, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 10226
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.