Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Perilipin Antibody, Novus Biologicals™
SDP

Catalog No. NB11040760T Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB11040760T 0.025 mL
NB11040760 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB11040760T Supplier Novus Biologicals Supplier No. NB11040760SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 7 publications

Perilipin Polyclonal specifically detects Perilipin in Human, Mouse, Rat, Porcine, Fish samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Imaging Mass Cytometry.

Specifications

Antigen Perilipin
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 2 ug/mL, Flow Cytometry 5 - 10 ug/mL, Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence reported in scientific literature, Immunohistochemistry-Paraffin 1:200, Imaging Mass Cytometry
Formulation Tris-Glycine and 0.15 M NaCl with 0.05% Sodium Azide
Gene Accession No. O60240
Gene Alias PERI, perilipin, perilipin 1, perilipin-1, PLINLipid droplet-associated protein
Gene Symbols PLIN1
Host Species Rabbit
Immunogen This Perilipin Antibody was developed against a synthetic peptide made to a region between residues 450-522 (C-terminus) of the human Perilipin protein. [Swiss-Prot# O60240]
Molecular Weight of Antigen 60 kDa
Purification Method Affinity Purified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Diabetes Research, Lipid and Metabolism, Lipid Droplets
Primary or Secondary Primary
Gene ID (Entrez) 5346
Target Species Human, Mouse, Rat, Pig, Fish
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.