Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Periostin/OSF-2 Antibody (OTI2B2), FITC, Novus Biologicals™
SDP

Catalog No. NBP273336F Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP273336F 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP273336F Supplier Novus Biologicals Supplier No. NBP273336F
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Periostin/OSF-2 Monoclonal specifically detects Periostin/OSF-2 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry.

Specifications

Antigen Periostin/OSF-2
Applications Immunohistochemistry
Classification Monoclonal
Clone OTI2B2
Conjugate FITC
Dilution Immunohistochemistry
Formulation PBS with 0.05% Sodium Azide
Gene Alias MGC119510, MGC119511, OSF-2osteoblast specific factor 2 (fasciclin I-like), OSF2periodontal ligament-specific periostin, Osteoblast-specific factor 2, PDLPOSTN, periostin, periostin isoform thy2, periostin isoform thy4, periostin isoform thy6, periostin isoform thy8, periostin, osteoblast specific factor, PNRP11-412K4.1
Gene Symbols POSTN
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 538-781 of human Periostin/OSF-2 (NP_001129407) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Cardiovascular Biology, Extracellular Matrix
Primary or Secondary Primary
Gene ID (Entrez) 10631.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.