Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Peripherin Antibody (7C5), Novus Biologicals™
SDP

Catalog No. NB300220 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB300220 0.5 mL
NB018150 0.025 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB300220 Supplier Novus Biologicals Supplier No. NB300220
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peripherin Monoclonal specifically detects Peripherin in Human (Negative), Mouse, Rat samples. It is validated for Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Western Blot.

Specifications

Antigen Peripherin
Applications Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 7C5
Concentration 1 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1:1000, Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence 1:500
Formulation 50% PBS, 50% glycerol with 5mM Sodium Azide
Gene Alias Neurofilament 4, neurofilament 4 (57kD), peripherin, PRPH1NEF4
Gene Symbols PRPH
Host Species Mouse
Immunogen Recombinant rat Peripherin purified from E. coli. [UniProt# P21807]
Molecular Weight of Antigen 57 kDa
Purification Method Tissue culture supernatant
Quantity 0.5 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5630
Test Specificity Chicken. Expected to cross-react with all mammals due to sequence homology.
Target Species Human (Negative), Mouse, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Supernatant
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.