Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Peripherin Antibody, Novus Biologicals™
SDP

Catalog No. NB300137 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB300137 0.1 mL
NB018134 0.025 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB300137 Supplier Novus Biologicals Supplier No. NB300137
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 6 publications

Peripherin Polyclonal specifically detects Peripherin in Human, Mouse, Rat, Porcine, Bovine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Specifications

Antigen Peripherin
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000-1:2000, Immunohistochemistry 1:2000, Immunocytochemistry/Immunofluorescence 1:2000, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Formulation Supplied as serum with 5mM Sodium Azide
Gene Alias Neurofilament 4, neurofilament 4 (57kD), peripherin, PRPH1NEF4
Gene Symbols PRPH
Host Species Rabbit
Immunogen Full length recombinant human peripherin expressed in and purified from E. coli.
Molecular Weight of Antigen 57 kDa
Purification Method Unpurified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cytoskeleton Markers, Neuronal Cell Markers, Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 5630
Test Specificity Specifically recognizes peripherin, showing no cross reactivity with other Class III intermediate filament proteins.
Target Species Human, Mouse, Rat, Pig, Bovine
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Antisera
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.