Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Peripherin Mouse anti-Rat, Mouse, Human, Clone: 7C5, MilliporeSigma™
Mouse Monoclonal Antibody
$188.90 - $485.00
Specifications
Antigen | Peripherin |
---|---|
Clone | 7C5 |
Concentration | 1.0 mg/mL |
Applications | Immunocytochemistry, Immunofluorescence, Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB5380I25
|
MilliporeSigma
MAB5380I25UG |
25 μg |
Each for $188.90
|
|
|||||
MAB5380I100
|
MilliporeSigma
MAB5380I100UG |
100 μg |
Each for $485.00
|
|
|||||
Description
Peripherin Monoclonal antibody specifically detects Peripherin in Rat, Mouse, Human samples. It is validated for Immunocytochemistry, Immunofluorescence, Western Blotting.Specifications
Peripherin | |
1.0 mg/mL | |
Monoclonal | |
Purified | |
RUO | |
Purified mouse monoclonal antibody IgG1 in PBS with 5 mM sodium azide with 50% glycerol. | |
IgG1 κ | |
Protein G purified |
7C5 | |
Immunocytochemistry, Immunofluorescence, Western Blot | |
Unconjugated | |
Mouse | |
Rat, Mouse, Human | |
Full length recombinant Rat peripherin expressed in and purified from E. coli. | |
Primary | |
Stable for 1 year at -10°C to -25°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title