Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PERK Antibody (5G5), Novus Biologicals™
SDP

Catalog No. NBP151661 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP151661 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP151661 Supplier Novus Biologicals Supplier No. NBP151661
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PERK Monoclonal specifically detects PERK in Human samples. It is validated for Western Blot, ELISA.

Specifications

Antigen PERK
Applications Western Blot, ELISA
Classification Monoclonal
Clone 5G5
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, ELISA 1:10000
Gene Alias DKFZp781H1925, EC 2.7.11.1, eukaryotic translation initiation factor 2 alpha kinase 3, eukaryotic translation initiation factor 2-alpha kinase 3, hsPEK, PEKPERKPancreatic eIF2-alpha kinase, PRKR-like endoplasmic reticulum kinase, WRS
Gene Symbols EIF2AK3
Host Species Mouse
Immunogen Purified recombinant fragment of human EIF2AK3 expressed in E. Coli.
Molecular Weight of Antigen 125 kDa
Purification Method Unpurified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 9451
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Ascites
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.