Learn More
Peroxidase, Horseradish, PROMO, Thermo Scientific Chemicals
Catalog No. J6002603PM
Click to view available options
Quantity:
100 mg
1 g
Used in the oxidation of luminol to 3-aminophthalate. It is often used in conjugates to determine the presence of a molecular target, especially in the detection of specific protein in a western blot after conjugation with antibody.
TRUSTED_SUSTAINABILITY
Specifications
Quantity 1 g
Form Lyophilized
RUO - Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.