Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Peroxiredoxin 3 Antibody (AT1F8), mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB332265 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB332265 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB332265 Supplier Novus Biologicals Supplier No. NBP312869MFV610

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Mouse Monoclonal Antibody

Peroxiredoxin 3 Monoclonal antibody specifically detects Peroxiredoxin 3 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence

Specifications

Antigen Peroxiredoxin 3
Applications Western Blot, ELISA, Immunocytochemistry
Classification Monoclonal
Clone AT1F8
Conjugate mFluor Violet 610 SE
Dilution Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias Antioxidant protein 1MGC104387, AOP-1MGC24293, AOP1PRO1748, EC 1.11.1.15, HBC189, MER5, peroxiredoxin 3, Peroxiredoxin III, peroxiredoxin-3, Protein MER5 homolog, prx-III, SP-22, thioredoxin-dependent peroxide reductase, mitochondrial
Host Species Mouse
Immunogen Recombinant human Peroxiredoxin3 (63-256aa) purified from E. coli (Accession number: NP_006784)
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 10935
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG3 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.