Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Peroxiredoxin 3 Mouse anti-Human, Clone: 1F8, Novus Biologicals™
SDP

Catalog No. NBP25942550 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP25942550 50 μL
NBP259425X 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP25942550 Supplier Novus Biologicals Supplier No. NBP25942550UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peroxiredoxin 3 Monoclonal specifically detects Peroxiredoxin 3 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Peroxiredoxin 3
Applications Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone 1F8
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000, Flow Cytometry 1:10-1:1035, ELISA, Immunocytochemistry/Immunofluorescence 1:100
Formulation Liquid. In PBS (pH 7.4), 10% Glycerol with 0.02% Sodium Azide
Gene Alias Antioxidant protein 1MGC104387, AOP-1MGC24293, AOP1PRO1748, EC 1.11.1.15, HBC189, MER5, peroxiredoxin 3, Peroxiredoxin III, peroxiredoxin-3, Protein MER5 homolog, prx-III, SP-22, thioredoxin-dependent peroxide reductase, mitochondrial
Gene Symbols PRDX3
Host Species Mouse
Immunogen Recombinant human Peroxiredoxin3 (63-256aa) purified from E. coli
Purification Method Protein A purified
Quantity 50 μL
Regulatory Status RUO
Research Discipline Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 10935
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG3 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.