Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Peroxiredoxin 4 Antibody (SAIC-40C-8), PerCP, Novus Biologicals™
SDP

Catalog No. NB340494 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB340494 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB340494 Supplier Novus Biologicals Supplier No. NBP320106PCP
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Peroxiredoxin 4 Monoclonal antibody specifically detects Peroxiredoxin 4 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/ Immunofluorescence, Mass Spectrometry

Specifications

Antigen Peroxiredoxin 4
Applications Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Mass Spectrometry
Classification Monoclonal
Clone SAIC-40C-8
Conjugate PerCP
Dilution Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/ Immunofluorescence, Mass Spectrometry
Formulation PBS
Gene Alias Antioxidant enzyme AOE372, AOE37-2PRX-4, EC 1.11.1.15, peroxiredoxin 4, Peroxiredoxin IV, peroxiredoxin-4, prx4, prx-4, Prx-IV, thioredoxin peroxidase (antioxidant enzyme), Thioredoxin peroxidase AO372, Thioredoxin-dependent peroxide reductase A0372
Host Species Rabbit
Immunogen Peptide LVQAFQYTDK derived from the cell redox-regulator Peroxiredoxin-4, conjugated to KLH.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 10549
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.