Learn More
Peroxiredoxin 4 Mouse anti-Human, Clone: CPTC-PRDX4-2, Novus Biologicals™
Shop All R&D Systems Products

Description
Specifications
Specifications
| Antigen | Peroxiredoxin 4 |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | CPTC-PRDX4-2 |
| Concentration | 0.2 mg/ml |
| Conjugate | Unconjugated |
| Dilution | ELISA |
| Formulation | 10 mM PBS with 0.05% BSA |
| Gene Alias | Antioxidant enzyme AOE372, AOE37-2PRX-4, EC 1.11.1.15, peroxiredoxin 4, Peroxiredoxin IV, peroxiredoxin-4, prx4, prx-4, Prx-IV, thioredoxin peroxidase (antioxidant enzyme), Thioredoxin peroxidase AO372, Thioredoxin-dependent peroxide reductase A0372 |
| Host Species | Mouse |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.