Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Peroxiredoxin 5 Rabbit anti-Human, Clone: 2, Novus Biologicals™

Rabbit Monoclonal Antibody
$353.50 - $560.50
Specifications
Antigen | Peroxiredoxin 5 |
---|---|
Clone | 2 |
Dilution | Western Blot 1:500-1:2000, Immunocytochemistry/Immunofluorescence 1:20-1:100, Immunoprecipitation 4-8 ul/mg of lysate, Knockout Validated |
Applications | Western Blot, Immunofluorescence, Immunoprecipitation, KnockDown |
Classification | Monoclonal |
Description
Peroxiredoxin 5 Monoclonal specifically detects Peroxiredoxin 5 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Knockout Validated.Specifications
Peroxiredoxin 5 | |
Western Blot 1:500-1:2000, Immunocytochemistry/Immunofluorescence 1:20-1:100, Immunoprecipitation 4-8 ul/mg of lysate, Knockout Validated | |
Monoclonal | |
Purified | |
RUO | |
0.2 um filtered solution in PBS | |
25824 | |
IgG | |
Protein A purified |
2 | |
Western Blot, Immunofluorescence, Immunoprecipitation, KnockDown | |
Unconjugated | |
Rabbit | |
Human | |
ACR1Peroxisomal antioxidant enzyme, Alu corepressor 1, Alu co-repressor 1, Antioxidant enzyme B166, AOEB166Peroxiredoxin V, B166, EC 1.11.1.15, Liver tissue 2D-page spot 71B, MGC117264, MGC142283, MGC142285, peroxiredoxin 5, peroxiredoxin-5, mitochondrial, PLPThioredoxin peroxidase PMP20, PMP20, PRDX6, prx-V, PRXV, SBBI10, Thioredoxin reductase, TPx type VI | |
Recombinant Human Peroxiredoxin 5 protein | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title