Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Peroxiredoxin 5 Rabbit anti-Human, Clone: 2, Novus Biologicals™
SDP

Catalog No. NB021191 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB021191 50 μg
NB021190 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB021191 Supplier Novus Biologicals Supplier No. NBP28986250UL
Only null left
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Peroxiredoxin 5 Monoclonal specifically detects Peroxiredoxin 5 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Knockout Validated.

Specifications

Antigen Peroxiredoxin 5
Applications Western Blot, Immunofluorescence, Immunoprecipitation, KnockDown
Classification Monoclonal
Clone 2
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, Immunocytochemistry/Immunofluorescence 1:20-1:100, Immunoprecipitation 4-8 ul/mg of lysate, Knockout Validated
Formulation 0.2 um filtered solution in PBS
Gene Alias ACR1Peroxisomal antioxidant enzyme, Alu corepressor 1, Alu co-repressor 1, Antioxidant enzyme B166, AOEB166Peroxiredoxin V, B166, EC 1.11.1.15, Liver tissue 2D-page spot 71B, MGC117264, MGC142283, MGC142285, peroxiredoxin 5, peroxiredoxin-5, mitochondrial, PLPThioredoxin peroxidase PMP20, PMP20, PRDX6, prx-V, PRXV, SBBI10, Thioredoxin reductase, TPx type VI
Host Species Rabbit
Immunogen Recombinant Human Peroxiredoxin 5 protein
Purification Method Protein A purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25824
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.