Learn More
Description
Specifications
Specifications
| Antigen | Peroxiredoxin 6 |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunofluorescence |
| Classification | Monoclonal |
| Clone | 220000000 |
| Conjugate | Alexa Fluor 405 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | 1-Cys, 24 kDa protein, Acidic calcium-independent phospholipase A2, aiPLA21-Cys PRX, Antioxidant protein 2, AOP2Non-selenium glutathione peroxidase, EC 1.11.1.15, EC 1.11.1.7, EC 3.1.1.-, KIAA0106Red blood cells page spot 12, Liver 2D page spot 40, MGC46173, NSGPx1-Cys peroxiredoxin, p29, peroxiredoxin 6, peroxiredoxin-6, PRX |
| Host Species | Mouse |
| Immunogen | Recombinant human Peroxiredoxin 6 (1-224aa) purified from E. coli |
| Purification Method | Protein A purified |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
