Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Peroxiredoxin 6 Mouse anti-Human, Mouse, Clone: 2.20E+08, Novus Biologicals™
SDP

Catalog No. NBP259436X Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP259436X 100 μL
NBP25943650 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP259436X Supplier Novus Biologicals Supplier No. NBP259436100UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peroxiredoxin 6 Monoclonal specifically detects Peroxiredoxin 6 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Peroxiredoxin 6
Applications Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone 220000000
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence
Formulation Liquid. In PBS (pH 7.4), 10% Glycerol with 0.02% Sodium Azide
Gene Alias 1-Cys, 24 kDa protein, Acidic calcium-independent phospholipase A2, aiPLA21-Cys PRX, Antioxidant protein 2, AOP2Non-selenium glutathione peroxidase, EC 1.11.1.15, EC 1.11.1.7, EC 3.1.1.-, KIAA0106Red blood cells page spot 12, Liver 2D page spot 40, MGC46173, NSGPx1-Cys peroxiredoxin, p29, peroxiredoxin 6, peroxiredoxin-6, PRX
Gene Symbols PRDX6
Host Species Mouse
Immunogen Recombinant human PRDX6 (1-224aa) purified from E. coli
Purification Method Protein A purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9588
Target Species Human, Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.