Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Peroxiredoxin, bacteria-like protein Antibody (V71) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP2596720.025ML
Description
Peroxiredoxin, bacteria-like protein Monoclonal specifically detects Peroxiredoxin, bacteria-like protein in Parasite samples. It is validated for Western Blot, ELISA, Immunohistochemistry.Specifications
Peroxiredoxin, bacteria-like protein | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:1, ELISA 1:1, Immunohistochemistry 1:1 | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Parasite | |
Purified |
Western Blot, ELISA, Immunohistochemistry | |
V71 | |
Unconjugated | |
PBS with 0.02% Sodium Azide | |
Peptide Sequence specific to peroxiredoxin antigen from Tetracapsuloides bryosalmonae (fish parasite). | |
0.025 mL | |
Virology Bacteria and Parasites | |
9682623 | |
Store at 4°C. Do not freeze. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction