Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Peroxiredoxin, bacteria-like protein Antibody (V71) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$196.10 - $499.50
Specifications
Antigen | Peroxiredoxin, bacteria-like protein |
---|---|
Clone | V71 |
Dilution | Western Blot 1:1, ELISA 1:1, Immunohistochemistry 1:1 |
Applications | Western Blot, ELISA, Immunohistochemistry |
Classification | Monoclonal |
Description
Peroxiredoxin, bacteria-like protein Monoclonal specifically detects Peroxiredoxin, bacteria-like protein in Parasite samples. It is validated for Western Blot, ELISA, Immunohistochemistry.Specifications
Peroxiredoxin, bacteria-like protein | |
Western Blot 1:1, ELISA 1:1, Immunohistochemistry 1:1 | |
Monoclonal | |
Purified | |
RUO | |
Parasite | |
9682623 | |
IgG | |
Protein A purified |
V71 | |
Western Blot, ELISA, Immunohistochemistry | |
Unconjugated | |
Mouse | |
Virology Bacteria and Parasites | |
PBS with 0.02% Sodium Azide | |
Peptide Sequence specific to peroxiredoxin antigen from Tetracapsuloides bryosalmonae (fish parasite). | |
Primary |
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title