Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Peroxiredoxin, bacteria-like protein Antibody (V71), mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB308789 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB308789 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB308789 Supplier Novus Biologicals Supplier No. NBP259672MFV450
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peroxiredoxin, bacteria-like protein Monoclonal antibody specifically detects Peroxiredoxin, bacteria-like protein in Parasite samples. It is validated for Western Blot, ELISA, Immunohistochemistry

Specifications

Antigen Peroxiredoxin, bacteria-like protein
Applications Western Blot, ELISA, Immunohistochemistry
Classification Monoclonal
Clone V71
Conjugate mFluor Violet 450 SE
Dilution Western Blot, ELISA, Immunohistochemistry
Formulation 50mM Sodium Borate
Host Species Mouse
Immunogen Peptide Sequence specific to peroxiredoxin antigen from Tetracapsuloides bryosalmonae (fish parasite).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Parasite
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.