Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PERP Antibody, mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB357504 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB357504 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB357504 Supplier Novus Biologicals Supplier No. NBP176741MFV500
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PERP Polyclonal antibody specifically detects PERP in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, Immunocytochemistry

Specifications

Antigen PERP
Applications Western Blot, ELISA, Immunocytochemistry, Immunocytochemistry
Classification Polyclonal
Conjugate mFluor Violet 500 SE
Dilution Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, Immunocytochemistry
Formulation 50mM Sodium Borate
Gene Alias dJ496H19.1, KCP1KCP-1, Keratinocyte-associated protein 1, KRTCAP11110017A08Rik, p53 apoptosis effector related to PMP22, p53 apoptosis effector related to PMP-22, P53-induced protein PIGPC1, PERP, TP53 apoptosis effector, PIGPC1RP3-496H19.1, THWkeratinocytes associated protein 1, Transmembrane protein THW
Host Species Rabbit
Immunogen Antibody was raised against a peptide corresponding to 17 amino acids near the carboxy terminus of human PERP. The immunogen is located within the last 50 amino acids of PERP. Amino Acid Sequence: EDDLLGNAKPRYFYTSA
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Checkpoint signaling, DNA Double Strand Break Repair, DNA Repair, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 64065
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.