Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PERP Antibody, Novus Biologicals™
SDP

Catalog No. NB432342 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
25ul
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB432342 25ul
NBP185173 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB432342 Supplier Novus Biologicals Supplier No. NBP18517325UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PERP Polyclonal specifically detects PERP in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen PERP
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000
Formulation PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Gene Alias dJ496H19.1, KCP1KCP-1, Keratinocyte-associated protein 1, KRTCAP11110017A08Rik, p53 apoptosis effector related to PMP22, p53 apoptosis effector related to PMP-22, P53-induced protein PIGPC1, PERP, TP53 apoptosis effector, PIGPC1RP3-496H19.1, THWkeratinocytes associated protein 1, Transmembrane protein THW
Gene Symbols PERP
Host Species Rabbit
Immunogen This antibody was developed against Recombinant Protein corresponding to amino acids:LQSSDHGQTSSLWWKCSQEGGGSGSYEEGCQSLMEYAW
Purification Method Affinity Purified
Quantity 25ul
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Checkpoint signaling, DNA Double Strand Break Repair, DNA Repair, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 64065
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.