Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PERQ2 Antibody, FITC, Novus Biologicals™
SDP

Catalog No. NB290479 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB290479 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB290479 Supplier Novus Biologicals Supplier No. NBP212813F
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PERQ2 Polyclonal antibody specifically detects PERQ2 in Human samples. It is validated for Western Blot, Immunoprecipitation

Specifications

Antigen PERQ2
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Conjugate FITC
Dilution Western Blot, Immunoprecipitation
Formulation PBS
Gene Alias DKFZp686J17223, FLJ23368, GRB10 interacting GYF protein 2, GRB10-interacting GYF protein 2, GYF domain containing 2, GYF2, KIAA0642DKFZp686I15154, PARK11, Parkinson disease (autosomal recessive, early onset) 11, PERQ amino acid rich, with GYF domain 2, PERQ amino acid rich, with GYF domain 3, PERQ amino acid-rich with GYF domain-containing protein 2, PERQ2, PERQ3, TNRC15, trinucleotide repeat containing 15, Trinucleotide repeat-containing gene 15 protein
Host Species Rabbit
Immunogen The immunogen this antibody was made to, maps to a region between residue 1249 and 1299 of human GRB10 Interacting GYF protein 2 using the numbering given in entry NP_001096616.1 (GeneID 26058).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26058
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.