Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EPA Method 8081 Pesticide Calibration Standard, SPEX CertiPrep™
SDP

Catalog No. 8081as Shop All Spex CertiPrep Products
Click to view available options
Quantity:
1 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

Includes

Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation

AnalyteCAS No.Concentration
1,2-Dibromo-3-chloropropane96-12-81000μg/mL
Chlorobenzilate510-15-61000μg/mL
Di-allate (mix of isomers)2303-16-41000μg/mL
Hexachlorobenzene118-74-11000μg/mL
Hexachlorocyclopentadiene77-47-41000μg/mL
Isodrin465-73-61000μg/mL
Kepone143-50-01000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Chemical Name or Material Pesticide Calibration Standard
Packaging 2mL amber prescored glass ampule
Concentration or Composition Notes Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity.
Methods For US EPA Methods 8080A, 8081A
Analyte or Component Names 1,2-Dibromo-3-chloropropane; Chlorobenzilate; Di-allate (mix of isomers); Hexachlorobenzene; Hexachlorocyclopentadiene; Isodrin; Kepone
For Use With (Equipment) GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation
Element 1,2-Dibromo-3-chloropropane, Chlorobenzilate, Di-allate (mix of isomers), Hexachlorobenzene, Hexachlorocyclopentadiene, Isodrin, Kepone
Solvent or Matrix Hexane:Toluene
Quantity 1 mL
Concentration or Composition (by Analyte or Components) 1000μg/mL in Hexane:Toluene

Product intended for laboratory use only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.