Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Petroleum Ether, puriss., Honeywell™ Riedel-de Haën™

Catalog No. p-7120661 Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
1 L
5 L
4 x 2.5 L
4 x 5 L
Packaging:
Aluminum Bottle
Glass Bottle

Catalog No. 60-045-646

Supplier: Honeywell Chemicals 245871L

Only null left
Add to Cart
Add to Cart

Puriss., low boiling point hydrogen treated naphtha, meets analytical specification of DAB, bpmin. 75% 40-60 °C (min. 75%)

  • Honeywell Riedel-de Haen has over 200 years of history in producing high-purity chemicals
  • It is the leading brand offering High-Purity Solvents for dedicated applications.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 101316-46-5
MDL Number MFCD00081849

Specifications

Density 0.642 to 0.656g/mL at 20°C
Quantity 1 L
Flash Point −40°C (−40°F)
Packaging Glass Bottle
UN Number UN1268
Grade puriss.
Chemical Name or Material Petroleum Ether, Meets analytical specification of DAB
Hazard Statement P210-P273-P301 + P310-P331-P391-P403 + P235
Precautionary Statement H225-H304-H336-H411
DOTInformation Class 3 - PG 2 - EHS - VP > 110kPa - Petroleum distillates, n.o.s., HI: S
EINECSNumber 309-852-0
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.