Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PEX11A Antibody, Novus Biologicals™
SDP

Catalog No. NBP15970520 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP15970520 20 μL
NBP159705 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP15970520 Supplier Novus Biologicals Supplier No. NBP15970520UL

Rabbit Polyclonal Antibody

PEX11A Polyclonal specifically detects PEX11A in Human samples. It is validated for Western Blot.

Specifications

Antigen PEX11A
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 0.2-1 ug/ml
Formulation PBS and 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. O75192
Gene Alias 28 kDa peroxisomal integral membrane protein, hsPEX11p, MGC119947, MGC138534, Peroxin-11A, peroxisomal biogenesis factor 11 alpha, Peroxisomal biogenesis factor 11Aperoxin-11A, peroxisomal membrane protein 11A, PEX11, PEX11-alpha, PMP28, Protein PEX11 homolog alpha
Gene Symbols PEX11A
Host Species Rabbit
Immunogen Synthetic peptides corresponding to PEX11A(peroxisomal biogenesis factor 11A) The peptide sequence was selected from the middle region of PEX11A (NP_003838). Peptide sequence MKRVTCDRAKKEKSASQDPLWFSVAEEETEWLQSFLLLLFRSLKQHPPLL.
Molecular Weight of Antigen 28 kDa
Purification Method Affinity Purified
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8800
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.