Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PEX14 Antibody, Novus Biologicals™
SDP

Catalog No. NBP171841 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP171841 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP171841 Supplier Novus Biologicals Supplier No. NBP171841
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PEX14 Polyclonal specifically detects PEX14 in Human samples. It is validated for Immunoprecipitation, Western Blot (Negative).

Specifications

Antigen PEX14
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Immunoprecipitation 2-5 ug/mg lysate, Western Blot (Negative)
Formulation Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
Gene Accession No. NP_004556.1
Gene Alias dJ734G22.2, MGC12767, NAPP2, NF-E2 associated polypeptide 2, peroxin-14, peroxisomal biogenesis factor 14, Peroxisomal membrane anchor protein PEX14, peroxisomal membrane anchor protein Pex14p, peroxisomal membrane protein PEX14, Pex14p, PTS1 receptor docking protein, PTS1 receptor-docking protein
Gene Symbols PEX14
Host Species Rabbit
Immunogen The immunogen for this product maps to a region between residue 250 and 300 of human Peroxisomal Biogenesis Factor 14 using the numbering given in entry NP_004556.1 (GeneID 5195).
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5195
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.