Learn More
PEX14 (Human Specific) Rabbit anti-Human, Unconjugated, Clone: 6K5, Proteintech

Rabbit Recombinant Antibody
$518.60
Specifications
| Antigen | PEX14 (Human Specific) |
|---|---|
| Clone | 6K5 |
| Applications | Western Blot, Immunofluorescence |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-225-6590
![]()
|
Proteintech Group Inc
801961RR100UL |
100 μL |
Pack of 1 for $518.60
|
|
|||||
Description
The PEX14 (human specific) antibody from Proteintech is a rabbit recombinant antibody generated with recombinant protein of human PEX14 (human specific). This antibody recognizes human antigen. The PEX14 (human specific) antibody has been validated for the following applications: IF, WB, ELISA analysis.
Specifications
| PEX14 (Human Specific) | |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| Human | |
| IgG |
| 6K5 | |
| Monoclonal | |
| Rabbit | |
| 5195 | |
| Primary |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.