Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PEX19 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA583740
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA583740 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA583740 Supplier Invitrogen™ Supplier No. PA583740
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SSMSEEELTK AMEGLGMDEG DGEGNILPIM QSIMQNLLSK DVLYPSLKEI TEKYPEWLQS HRESLP Highest antigen sequence indentity to the following orthologs: Mouse - 89%, Rat - 92%.

This gene is necessary for early peroxisomal biogenesis. It acts both as a cytosolic chaperone and as an import receptor for peroxisomal membrane proteins (PMPs). Peroxins (PEXs) are proteins that are essential for the assembly of functional peroxisomes. The peroxisome biogenesis disorders (PBDs) are a group of genetically heterogeneous autosomal recessive, lethal diseases characterized by multiple defects in peroxisome function. The peroxisomal biogenesis disorders are a heterogeneous group with at least 14 complementation groups and with more than 1 phenotype being observed in cases falling into particular complementation groups. Although the clinical features of PBD patients vary, cells from all PBD patients exhibit a defect in the import of one or more classes of peroxisomal matrix proteins into the organelle. Defects in this gene are a cause Zellweger syndrome (ZWS).
TRUSTED_SUSTAINABILITY

Specifications

Antigen PEX19
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PEX19
Gene Accession No. P40855, Q8VCI5, Q9QYU1
Gene Alias 33 kDa housekeeping protein; D1S2223E; HK33; housekeeping gene, 33kD; OK/SW-cl.22; PBD12A; peroxin-19; Peroxisomal biogenesis factor 19; peroxisomal farnesylated protein; peroxisome biogenesis factor 19; Pex19; PMP1; PMPI; PxF; PXMP1
Gene Symbols PEX19
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PEX19. Recombinant protein control fragment (Product #RP-100414).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19298, 289233, 5824
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.