Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PEX26 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP15511420UL
Description
PEX26 Polyclonal specifically detects PEX26 in Human samples. It is validated for Western Blot.Specifications
PEX26 | |
Polyclonal | |
Western Blot 1:100-1:2000 | |
Q7Z412 | |
PEX26 | |
Synthetic peptides corresponding to PEX26(peroxisome biogenesis factor 26) The peptide sequence was selected from the N terminal of PEX26. Peptide sequence MKSDSSTSAAPLRGLGGPLRSSEPVRAVPARAPAVDLLEEAADLLVVHLD. | |
Affinity Purified | |
RUO | |
55670 | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
FLJ20695, peroxin-26, peroxisomal biogenesis factor 26, peroxisome assembly protein 26, peroxisome biogenesis disorder, complementation group 8, peroxisome biogenesis disorder, complementation group A, peroxisome biogenesis factor 26, PEX26M1T, Pex26pM1T | |
Rabbit | |
34 kDa | |
20 μL | |
Primary | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction