Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PEX26 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00 - $487.50
Specifications
Antigen | PEX26 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP1551420
![]() |
Novus Biologicals
NBP15511420UL |
20 μL |
Each for $206.00
|
|
|||||
NBP155114
![]() |
Novus Biologicals
NBP155114 |
100 μL |
Each for $487.50
|
|
|||||
Description
PEX26 Polyclonal specifically detects PEX26 in Human samples. It is validated for Western Blot.Specifications
PEX26 | |
Polyclonal | |
Rabbit | |
Q7Z412 | |
55670 | |
Synthetic peptides corresponding to PEX26(peroxisome biogenesis factor 26) The peptide sequence was selected from the N terminal of PEX26. Peptide sequence MKSDSSTSAAPLRGLGGPLRSSEPVRAVPARAPAVDLLEEAADLLVVHLD. | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
FLJ20695, peroxin-26, peroxisomal biogenesis factor 26, peroxisome assembly protein 26, peroxisome biogenesis disorder, complementation group 8, peroxisome biogenesis disorder, complementation group A, peroxisome biogenesis factor 26, PEX26M1T, Pex26pM1T | |
PEX26 | |
IgG | |
34 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title