Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PEX7 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00 - $487.50
Specifications
Antigen | PEX7 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP15757820
![]() |
Novus Biologicals
NBP15757820UL |
20 μL |
Each for $206.00
|
|
|||||
NBP157578
![]() |
Novus Biologicals
NBP157578 |
100 μL |
Each for $487.50
|
|
|||||
Description
PEX7 Polyclonal specifically detects PEX7 in Human samples. It is validated for Western Blot.Specifications
PEX7 | |
Polyclonal | |
Rabbit | |
O00628 | |
5191 | |
Synthetic peptides corresponding to PEX7(peroxisomal biogenesis factor 7) The peptide sequence was selected from the N terminal of PEX7. Peptide sequence MSAVCGGAARMLRTPGRHGYAAEFSPYLPGRLACATAQHYGIAGCGTLLI. | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
peroxin-7, peroxisomal biogenesis factor 7, peroxisomal targeting signal 2 receptor, peroxisome targeting signal 2 receptor, PTS2 receptor, PTS2Rperoxisomal PTS2 receptor, RCDP1, RD | |
PEX7 | |
IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title