Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Pexidartinib
GREENER_CHOICE

Catalog No. p-7224938
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Potent CSF-1R inhibitor

Pexidartinib is a potent colony-stimulating factor-1 receptor (CSF-1R) tyrosine kinase inhibitor (IC50 = 20 nM). In the B16F10 mouse melanoma model, Pexidartinib reduces the systemic and local accumulation of macrophages without affecting Gr-1+ myeloid derived suppressor cells. When combined with tumor-specific CD8 T cell immunotherapy, Pexidartinib promotes the control of tumor growth. In the BRAFV600E-driven mouse melanoma model, Pexidartinib improves the efficacy of adoptive cell therapy with reduction of TIMs and an increase in tumor-infiltrating lymphocytes and T cells. Orally bioavailable.

Specifications

Chemical Name or Material N-[5-[(5-Chloro-1 H-pyrrolo[2,3-b]pyridin-3-yl)methyl]-2-pyridinyl]-6-(trifluoromethyl)-3-pyridinemethanamine
CAS 1029044-16-3
Quantity 10 mg
Target Additional RTK Inhibitors
Molecular Formula C20H15ClF3N5
Purity 98%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.