Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ PF 04449613, >98%
GREENER_CHOICESDP

Catalog No. 591510 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
59-151-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 59-151-0 Supplier Tocris Bioscience™ Supplier No. 5915/10

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Potent PDE9 inhibitor; brain penetrant

PF 04449613 is a potent PDE9 inhibitor (IC50 = 22 nM). Also exhibits a high affinity for cGMP (Km ∼170 nM). PF 04449613 reduces body fat in mice with diet-induced obesity, stimulating mitochondrial activity in brown and white fat, and improving cardiometabolic syndrome symptoms. Brain penetrant.

Chemical Identifiers

CAS 1236858-52-8
Molecular Formula C21H25N5O3

Specifications

Quantity 10 mg
Solubility Soluble to 100mM in DMSO and to 10mM in 1eq. HCl with gentle warming
Formula Weight 395.45
Purity >98%
Molecular Weight (g/mol) 395.463
Chemical Name or Material 1,5-Dihydro-6-[(1R)-1-(3-phenoxy-1-azetidinyl)ethyl]-1-(tetrahydro-2H-pyran-4-yl)-4H-pyrazolo[3,4-d]
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.