Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ PF 04449913 maleate

Click to view available options
Quantity:
5 mg
25 mg
Description
PF 04449913 maleate is a potent Smo antagonist (IC50 = 5 nM). Attenuates the leukemia-initiation potential of AML cells in a serial transplantation mouse model. Also eliminates self-propagation capacity of AML cells.
Specifications
Specifications
| Chemical Name or Material | 1-((2R,4R)-2-(1H-Benzimidazol-2-yl)-1-methylpiperidin-4-yl)-3-(4-cyanophenyl)urea maleate salt |
| CAS | 2030410-25-2 |
| Synonym | glasdegib, PF-913 |
| Quantity | 5 mg |
| Molecular Formula | C21H22N6O.C4H4O4 |
| Purity | 98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction