Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ PF 04620110
GREENER_CHOICE

Catalog No. p-7116085
Click to view available options
Quantity:
5 mg
25 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Potent and selective DGAT-1 inhibitor; orally bioavailable

PF 04620110 is a potent and selective DGAT-1 inhibitor (IC50 = 19 nM). Exhibits >1000-fold selectivity for DGAT-1 over DGAT-2, and no significant activity against a range of other enzymes, ion channels and receptors. Reduces plasma triglyceride levels in a rat lipid challenge model. Orally bioavailable.

Specifications

Chemical Name or Material trans-4-[4-(4-Amino-7,8-dihydro-5-oxopyrimido[5,4-f][1,4]oxazepin-6(5H)-yl)phenyl]cyclohexaneacetic acid
CAS 1109276-89-2
Assay Percent Range >98%
Quantity 25 mg
Molecular Formula C21H24N4O4
Formula Weight Observed MW: 396.44
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.