Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ PF 04628935
GREENER_CHOICE

Catalog No. p-7116127
Click to view available options
Quantity:
5 mg
25 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Potent ghrelin receptor inverse agonist

PF 04628935 is a potent ghrelin receptor inverse agonist (IC50 = 4.6 nM). Orally bioavailable and exhibits reasonable brain penetration.

Specifications

Chemical Name or Material 1-[2-[[2-Chloro-4-(2H-1,2,3-triazol-2-yl)phenyl]methyl]-2,7-diazaspiro[3.5]non-7-yl]-2-(2-methylimidazo[2,1-b]thiazol-6-yl)ethanone
CAS 1383719-97-8
Assay Percent Range >98%
Quantity 25 mg
Molecular Formula C24H26ClN7OS
Formula Weight Observed MW: 496.03
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.