Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ PF 06465469
GREENER_CHOICESDP

Catalog No. 471010 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
47-101-0 10 mg
47-105-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 47-101-0 Supplier Tocris Bioscience™ Supplier No. 4710/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent ITK inhibitor; also inhibits BTK

Potent inhibitor of interleukin-2 inducible T cell kinase (ITK) (IC50 = 2 nM). Also exhibits inhibitory activity against Bruton's tyrosine kinase (BTK) (IC50 = 2 nM). Displays nanomolar potencies in a cell-based IP1 assay and human whole blood assay (IC50 values are 31 and 48 nM respectively).

Chemical Identifiers

CAS 1407966-77-1
Molecular Formula C30H33N7O2

Specifications

Quantity 10 mg
Formula Weight 523.63
Purity >95%
Molecular Weight (g/mol) 523.641
Chemical Name or Material PF 06465469
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.