Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ PF 3845
GREENER_CHOICE

Catalog No. 417510
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
41-751-0 10 mg
41-755-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 41-751-0 Supplier Tocris Bioscience™ Supplier No. 4175/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Selective FAAH inhibitor

Selective fatty acid amide hydrolase (FAAH) inhibitor (Ki = 0.23 μM). Reduces inflammatory pain via a cannabinoid receptor-dependent mechanism. Highly efficacious and selective in vivo. Displays no activity at FAAH-2 (IC50 >10 μM).

Chemical Identifiers

CAS 1196109-52-0
Molecular Formula C24H23F3N4O2

Specifications

Quantity 10 mg
Formula Weight 456.46
Purity >99%
Molecular Weight (g/mol) 456.47
Chemical Name or Material PF 3845
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.