Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ PF 429242
GREENER_CHOICE

Catalog No. 335410
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
33-541-0 10 mg
33-545-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 33-541-0 Supplier Tocris Bioscience™ Supplier No. 3354/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Competitive inhibitor of SREBP site 1 protease

Reversible, competitive inhibitor of sterol regulatory element-binding protein (SREBP) site 1 protease (IC50 = 0.175 μM). Selective for site 1 protease against a panel of serine proteases. Inhibits rate of cholesterol synthesis in CHO cells (IC50 = 0.53 μM). Also displays antiviral activity. Cell permeable.

Chemical Identifiers

CAS 947303-87-9
Molecular Formula C25H35N3O2

Specifications

Quantity 10 mg
Formula Weight 482.49
Purity >97%
Molecular Weight (g/mol) 409.574
Chemical Name or Material PF 429242
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.