Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PF1 Antibody, Novus Biologicals™
SDP

Catalog No. NB10081671 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10081671 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10081671 Supplier Novus Biologicals Supplier No. NB10081671
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 2 publications

PF1 Polyclonal specifically detects PF1 in Chicken, Human, Mouse samples. It is validated for Chemotaxis, ChIP Assay, Immunoprecipitation, Western Blot.

Specifications

Antigen PF1
Applications Chemotaxis, ChIP Assay, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:2000-1:10000, Immunoprecipitation 2-5 ug/mg lysate, Chromatin Immunoprecipitation (ChIP)
Formulation Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
Gene Accession No. Q96QT6
Gene Alias FLJ34122, KIAA1523MGC131914, PF1, PHD factor 1, PHD finger protein 12, PHD zinc finger transcription factor
Gene Symbols PHF12
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 954 and 1004 of human PHD factor 1 using the numbering given in entry NP_001028733.1 (GeneID 57649).
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57649
Target Species Chicken, Human, Mouse
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.