Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PF1 Polyclonal Antibody, Bethyl Laboratories

Catalog No. 501560899
Encompass_Preferred
Change view
Click to view available options
Quantity:
20 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-0899 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-0899 Supplier Bethyl Laboratories, Inc Supplier No. A301647AT
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

Acts as a transcriptional repressor. Involved in recruitment of functional SIN3A complexes to DNA. Represses transcription at least in part through the activity of an associated histone deacetylase (HDAC). May also repress transcription in a SIN3A-independent manner through recruitment of functional AES complexes to DNA.

Specifications

Antigen PF1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene PHF12
Gene Accession No. Q5SPL2, Q96QT6
Gene Alias PHD finger protein 12, PHD factor 1, Pf1, PHF12 (HGNC:20816)
Gene Symbols Phf12
Host Species Rabbit
Immunogen Between 954 and 1004
Purification Method Antigen affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 268448, 57649
Target Species Human, Mouse
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.