Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PFDN5 Antibody, mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB332394 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB332394 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB332394 Supplier Novus Biologicals Supplier No. NBP297061MFV450
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PFDN5 Polyclonal antibody specifically detects PFDN5 in Human samples. It is validated for ELISA

Specifications

Antigen PFDN5
Applications ELISA
Classification Polyclonal
Conjugate mFluor Violet 450 SE
Dilution ELISA
Formulation 50mM Sodium Borate
Gene Alias c-myc binding protein, C-Myc-binding protein Mm-1, MM-1, MM1MGC71907, Myc modulator 1, myc modulator-1, PFD5MGC5329, prefoldin 5, prefoldin subunit 5
Host Species Rabbit
Immunogen Rabbits immunized with purified, recombinant Human PFDN5 (Accession#: NP_002615.2, Met1-Ala154)
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 5204
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.